missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methylumbelliferyl-α-D-galactopyranoside hydrate, 99%, ACROS Organics™

Chemical Identifiers

CAS 38597-12-5
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C16H18O8·xH2O
Formula Weight 338.31
MDL Number MFCD03791284
Synonym 4-μ-α-D-gal
Chemical Name or Material 4-Methylumbelliferyl-α-D-galactopyranoside hydrate, 99%
Assay ≥98.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC337162500 SDS Thermo Fisher Scientific
AC337162500
Glass bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

38597-12-5
100.0
C16H18O8·xH2O
MFCD03791284
4-Methylumbelliferyl-α-D-galactopyranoside hydrate, 99%
98.5
99%
338.31
4-μ-α-D-gal
≥98.5%

Specifications

Crystalline Powder
250mg
100.0
4-μ-α-D-gal
98.5
Glass bottle
C16H18O8·xH2O
38597-12-5
≥98.5%
338.31
White to almost white
8% max. (K.F.)
(5% soln. in DMF) clear, practically colorless
Authentic
99%
MFCD03791284
4-Methylumbelliferyl-α-D-galactopyranoside hydrate, 99%