Please login to your online account to display your discounted pricing

4-Methylumbelliferyl-β-D-galactopyranoside, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  AC337210050

 View more versions of this product

Catalog No. AC337210050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder
Formula Weight 338.31
Quantity 5g
Color White to slightly off-white
EINECS Number 228-185-5
Assay Complies
Specific Rotation Condition −64° (20°C c=1,pyridine,24h)
Beilstein 18, IV, 339
Chemical Name or Material 4-Methylumbelliferyl-β-D-galactopyranoside
Infrared Spectrum Authentic
Packaging Glass bottle
Specific Rotation -62° to -66° (20°C, 589nm) (c=1, pyridine, after 24h)
Molecular Formula C16H18O8
MDL Number MFCD00036773
CAS 6160-78-7
Health Hazard 1 MOISTURE SENSITIVE
Appearance of Solution (0.5% in DMF) clear colorless; (1% in pyridine) clear colorless

Specifications

Chemical Name or Material 4-Methylumbelliferyl-β-D-galactopyranoside
Assay Complies
Molecular Formula C16H18O8
Formula Weight 338.31
MDL Number MFCD00036773
Accessories

MOISTURE SENSITIVE

SDS & Safety Documents