missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Methylumbelliferyl-β-D-galactopyranoside, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 337210010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Methylumbelliferyl-β-D-galactopyranoside | |
248°C | |
Authentic | |
C16H18O8 | |
1 g | |
−64° (20°C c=1,pyridine,24h) | |
4-methylumbelliferyl-beta-d-galactopyranoside, 4-methylumbelliferyl-galactopyranoside, 4-methylumbelliferyl beta-d-galactoside, 4-methylumbelliferyl b-d-galactoside, muga, 4-methylumbelliferyl beta-galactoside, 7-beta-d-galactopyranosyloxy-4-methyl-2h-1-benzopyran-2-one, 4-methylumbelliferyl b-d-galactopyranoside, 4-methyl-7-2s,3r,4s,5r,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxy-chromen-2-one, 4-methyl-7-2s,3r,4s,5r,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy chromen-2-one | |
YUDPTGPSBJVHCN-DZQJYWQESA-N | |
4-methyl-7-[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-2-one | |
93577 | |
Complies |
6160-78-7 | |
White | |
Glass bottle | |
MFCD00036773,MFCD00063694 | |
18,V,1,444 | |
-64 | |
Solubility in water: moderately soluble. Other solubilities: soluble in dmf,pyridine,moderately soluble in ethanol,dmso | |
CC1=CC(=O)OC2=CC(O[C@@H]3O[C@H](CO)[C@H](O)[C@H](O)[C@H]3O)=CC=C12 | |
338.31 | |
338.31 | |
Powder |
Chemical Identifiers
6160-78-7 | |
338.31 | |
YUDPTGPSBJVHCN-DZQJYWQESA-N | |
93577 | |
CC1=CC(=O)OC2=CC(O[C@@H]3O[C@H](CO)[C@H](O)[C@H](O)[C@H]3O)=CC=C12 |
C16H18O8 | |
MFCD00036773,MFCD00063694 | |
4-methylumbelliferyl-beta-d-galactopyranoside, 4-methylumbelliferyl-galactopyranoside, 4-methylumbelliferyl beta-d-galactoside, 4-methylumbelliferyl b-d-galactoside, muga, 4-methylumbelliferyl beta-galactoside, 7-beta-d-galactopyranosyloxy-4-methyl-2h-1-benzopyran-2-one, 4-methylumbelliferyl b-d-galactopyranoside, 4-methyl-7-2s,3r,4s,5r,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxy-chromen-2-one, 4-methyl-7-2s,3r,4s,5r,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy chromen-2-one | |
4-methyl-7-[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-2-one |
Safety and Handling
MOISTURE SENSITIVE
EINECSNumber : 228-185-5
RUO – Research Use Only