missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methylumbelliferyl beta-D-Ribofuranoside, TCI America™

$305.56 - $918.22

Chemical Identifiers

CAS 195385-93-4
Molecular Formula C15H16O7
Molecular Weight (g/mol) 308.286
MDL Number MFCD06796941
InChI Key FAGLTVBWEMHJRP-NMFUWQPSSA-N
PubChem CID 18731513
IUPAC Name 7-[(2S,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]oxy-4-methylchromen-2-one
SMILES CC1=CC(=O)OC2=C1C=CC(=C2)OC3C(C(C(O3)CO)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M3027100MG View Documents TCI America
M3027100MG
100mg Each for $918.22
 
M302725MG View Documents TCI America
M302725MG
25mg Each for $305.56
 
Specifications

Chemical Identifiers

195385-93-4
308.286
FAGLTVBWEMHJRP-NMFUWQPSSA-N
7-[(2S,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]oxy-4-methylchromen-2-one
C15H16O7
MFCD06796941
18731513
CC1=CC(=O)OC2=C1C=CC(=C2)OC3C(C(C(O3)CO)O)O

Specifications

White
195385-93-4
MFCD06796941
CC1=CC(=O)OC2=C1C=CC(=C2)OC3C(C(C(O3)CO)O)O
308.286
308.29
≥98.0% (HPLC)
100mg
C15H16O7
FAGLTVBWEMHJRP-NMFUWQPSSA-N
7-[(2S,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]oxy-4-methylchromen-2-one
18731513
Crystal-Powder at 20°C
4-Methylumbelliferyl beta-D-Ribofuranoside
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer (-20 °C)