missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methylumbelliferyl beta-D-glucopyranoside, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals J66630MD

 View more versions of this product

Catalog No. AAJ66630MD



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Methylumbelliferyl beta-D-glucopyranoside
210°C to 212°C
C16H18O8
250 mg
4-methylumbelliferyl, a-d-glucoside, glc1-b-4mu, 7-, a-d-glucopyranosyloxy-4-methyl-2h-1-benzopyran-2-one, 4-methylumbelliferyl glucopyranoside, 4-methyl-7-2s,4s,5s-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy chromen-2-one
YUDPTGPSBJVHCN-YMILTQATSA-N
4-methyl-7-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-2-one
2733779
99%
18997-57-4
White
MFCD00036773
94673
Soluble in DMF or water
CC1=CC(=O)OC2=C1C=CC(=C2)OC3C(C(C(C(O3)CO)O)O)O
338.312
338.31
Powder

Chemical Identifiers

18997-57-4
338.312
YUDPTGPSBJVHCN-YMILTQATSA-N
2733779
CC1=CC(=O)OC2=C1C=CC(=C2)OC3C(C(C(C(O3)CO)O)O)O
C16H18O8
MFCD00036773
4-methylumbelliferyl, a-d-glucoside, glc1-b-4mu, 7-, a-d-glucopyranosyloxy-4-methyl-2h-1-benzopyran-2-one, 4-methylumbelliferyl glucopyranoside, 4-methyl-7-2s,4s,5s-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy chromen-2-one
4-methyl-7-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-2-one
Safety and Handling

Safety and Handling

EINECSNumber : 242-736-7

TSCA : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only