missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methylumbelliferyl alpha-D-Mannopyranoside, TCI America™

$160.42 - $481.27

Chemical Identifiers

CAS 28541-83-5
Molecular Formula C16H18O8
Molecular Weight (g/mol) 338.312
MDL Number MFCD00067662
InChI Key YUDPTGPSBJVHCN-VMMWWAARSA-N
PubChem CID 2733782
IUPAC Name 4-methyl-7-[(2R,3S,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-2-one
SMILES CC1=CC(=O)OC2=C1C=CC(=C2)OC3C(C(C(C(O3)CO)O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M302325MG View Documents TCI America
M302325MG
25mg Each for $160.42
 
M3023100MG View Documents TCI America
M3023100MG
100mg Each for $481.27
 
Specifications

Chemical Identifiers

28541-83-5
338.312
YUDPTGPSBJVHCN-VMMWWAARSA-N
4-methyl-7-[(2R,3S,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-2-one
C16H18O8
MFCD00067662
2733782
CC1=CC(=O)OC2=C1C=CC(=C2)OC3C(C(C(C(O3)CO)O)O)O

Specifications

White-Yellow
25mg
C16H18O8
YUDPTGPSBJVHCN-VMMWWAARSA-N
4-methyl-7-[(2R,3S,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-2-one
2733782
Crystal-Powder at 20°C
4-Methylumbelliferyl alpha-D-Mannopyranoside
217°C
28541-83-5
MFCD00067662
CC1=CC(=O)OC2=C1C=CC(=C2)OC3C(C(C(C(O3)CO)O)O)O
338.312
338.31
≥97.0% (HPLC)
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer (-20 °C)