missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methylumbelliferyl acetate, 99%

Manufacturer:  Alfa Aesar™ A1214714

 View more versions of this product

Catalog No. AAA1214714



Specifications

Specifications

4-Methylumbelliferyl acetate
99%
MFCD00006865
7-Acetoxy-4-methylcoumarin
CC1=CC(=O)OC2=C1C=CC(=C2)OC(=O)C
218.21
CHEBI:17763
2747-05-9
C12H10O4
189667
HXVZGASCDAGAPS-UHFFFAOYSA-N
(4-methyl-2-oxochromen-7-yl) acetate
366
218.21

Chemical Identifiers

2747-05-9
218.21
HXVZGASCDAGAPS-UHFFFAOYSA-N
366
(4-methyl-2-oxochromen-7-yl) acetate
C12H10O4
MFCD00006865
7-Acetoxy-4-methylcoumarin
CHEBI:17763
CC1=CC(=O)OC2=C1C=CC(=C2)OC(=O)C
Safety and Handling

Safety and Handling

Recommended Storage : 2°C to 8°C

SDS Documents