missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Methylumbelliferone 98.0+%, TCI America™
Supplier: TCI America M0453500G
Specifications
4-Methylumbelliferone | |
188°C | |
C10H8O3 | |
500 g | |
HSHNITRMYYLLCV-UHFFFAOYSA-N | |
7-hydroxy-4-methyl-2H-chromen-2-one | |
5280567 | |
176.17 | |
Crystalline Powder |
90-33-5 | |
White-Yellow | |
MFCD00006866 | |
4-methylumbelliferone, hymecromone, 7-hydroxy-4-methylcoumarin, imecromone, 7-hydroxy-4-methyl-2h-chromen-2-one, cholestil, mendiaxon, 4-methyl-7-hydroxycoumarin, beta-methylumbelliferone, bilcolic | |
CC1=CC(=O)OC2=CC(O)=CC=C12 | |
176.17 | |
CHEBI:17224 | |
≥98.0% (T) |
Chemical Identifiers
90-33-5 | |
176.17 | |
HSHNITRMYYLLCV-UHFFFAOYSA-N | |
5280567 | |
7-hydroxy-4-methyl-2H-chromen-2-one |
C10H8O3 | |
MFCD00006866 | |
4-methylumbelliferone, hymecromone, 7-hydroxy-4-methylcoumarin, imecromone, 7-hydroxy-4-methyl-2h-chromen-2-one, cholestil, mendiaxon, 4-methyl-7-hydroxycoumarin, beta-methylumbelliferone, bilcolic | |
CHEBI:17224 | |
CC1=CC(=O)OC2=CC(O)=CC=C12 |
Safety and Handling
EINECSNumber : (9)-1048
RTECSNumber : GN7000000
TSCA : Yes