missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methyltriphenylamine, TCI America™

$180.28 - $534.73

Chemical Identifiers

CAS 4316-53-4
Molecular Formula C19H17N
Molecular Weight (g/mol) 259.352
InChI Key IULUNTXBHHKFFR-UHFFFAOYSA-N
Synonym N,N-Diphenyl-p-toluidine
PubChem CID 11299928
IUPAC Name 4-methyl-N,N-diphenylaniline
SMILES CC1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=CC=C3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M18985G View Documents TCI America
M18985G
5g Each for $534.73
 
M18981G View Documents TCI America
M18981G
1g Each for $180.28
 
Specifications

Chemical Identifiers

4316-53-4
259.352
N,N-Diphenyl-p-toluidine
4-methyl-N,N-diphenylaniline
C19H17N
IULUNTXBHHKFFR-UHFFFAOYSA-N
11299928
CC1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=CC=C3

Specifications

White-Yellow
5g
C19H17N
IULUNTXBHHKFFR-UHFFFAOYSA-N
4-methyl-N,N-diphenylaniline
11299928
Crystal-Powder at 20°C
4-Methyltriphenylamine
67°C
4316-53-4
N,N-Diphenyl-p-toluidine
CC1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=CC=C3
259.352
259.35
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No