Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methylthiomorpholine 1,1-dioxide, 97%

$66.15 - $232.04

Chemical Identifiers

Chemical Name or Material 4-Methylthiomorpholine 1,1-dioxide
CAS 25343-91-3
Molecular Formula C5H11NO2S
Formula Weight 149.21
MDL Number MFCD00014623
Beilstein 114527
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0223503

  SDS
alfa aesar™
L0223503
1g Each for $66.15

AAL0223506

  SDS
alfa aesar™
L0223506
5g Each for $232.04
Description & Specifications

Specifications

Formula Weight 149.21
CAS 25343-91-3
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
Health Hazard 1 Warning
MDL Number MFCD00014623
Molecular Formula C5H11NO2S
Quantity 1g
Beilstein 114527
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
RTECS Number XM3015000
Melting Point 84° to 88°C
Chemical Name or Material 4-Methylthiomorpholine 1,1-dioxide

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.