Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Methylthio)thiophenol, 96%

$61.64 - $207.93

Chemical Identifiers

Chemical Name or Material 4-(Methylthio)thiophenol
CAS 1122-97-0
Molecular Formula C7H8S2
Formula Weight 156.27
MDL Number MFCD00082770
Beilstein 1931500
Synonym 4-(Methylmercapto)thiophenol; 4-(Methylthio)benzenethiol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0958003

  SDS
alfa aesar™
L0958003
1g Each for $61.64

AAL0958006

  SDS
alfa aesar™
L0958006
5g Each for $207.93
Description & Specifications

Specifications

Formula Weight 156.27
Chemical Name or Material 4-(Methylthio)thiophenol
Assay 96%
Molecular Formula C7H8S2
MDL Number MFCD00082770
Beilstein 1931500
Synonym 4-(Methylmercapto)thiophenol; 4-(Methylthio)benzenethiol
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
CAS 1122-97-0
Melting Point 12° to 16°C
Boiling Point 116° to 117°C (3mmHg)
Quantity 1g
Health Hazard 3 GHS P Statement
P280-P301+P310-P305+P351+P338-P362-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P280-P301+P310-P305+P351+P338-P362-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.