missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Methylthio)thiophenol, 96%

Manufacturer:  Alfa Aesar™ L0958003

 View more versions of this product

Catalog No. AAL0958003



Specifications

Specifications

4-(Methylthio)thiophenol
12°C to 16°C
1122-97-0
MFCD00082770
4-methylthio thiophenol, 4-methylthio benzenethiol, 4-methylthiothiophenol, 4-methylthio thiophenol, 4-methylsulfanyl benzenethiol, 4-methylsulfanyl thiophenol, p-methylthiothiophenol, 4-mercaptothioanisole, pubchem6825, 4-methylthio-thiophenol
CSC1=CC=C(C=C1)S
156.261
156.27
116°C to 117°C (3mmHg)
1g
C7H8S2
1931500
KYMOWQQIZINTJZ-UHFFFAOYSA-N
4-methylsulfanylbenzenethiol
136896
96%

Chemical Identifiers

1122-97-0
156.261
KYMOWQQIZINTJZ-UHFFFAOYSA-N
136896
CSC1=CC=C(C=C1)S
C7H8S2
MFCD00082770
4-methylthio thiophenol, 4-methylthio benzenethiol, 4-methylthiothiophenol, 4-methylthio thiophenol, 4-methylsulfanyl benzenethiol, 4-methylsulfanyl thiophenol, p-methylthiothiophenol, 4-mercaptothioanisole, pubchem6825, 4-methylthio-thiophenol
4-methylsulfanylbenzenethiol
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P301+P310-P305+P351+P338-P362-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Documents