missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Methylthio)phenylzinc iodide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles

Chemical Identifiers

CAS 307496-24-8
Formula Weight 315.48
MDL Number MFCD01863641
UN Number UN3399
Chemical Name or Material 4-(Methylthio)phenylzinc iodide
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH58137AD SDS Alfa Aesar™
H58137AD
50mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

  • Packaged under Argon
Description & Specifications

Chemical Identifiers

307496-24-8
MFCD01863641
4-(Methylthio)phenylzinc iodide
315.48
UN3399

Specifications

315.48
UN3399
Air Sensitive
50mL
MFCD01863641
4-(Methylthio)phenylzinc iodide
ChemSeal™ bottles
307496-24-8
Yellow to brown to black
Liquid
0.5M in THF

Safety and Handling

GHS H Statement
H225-H261-H314-H351-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.

GHS P Statement
P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Handle under inert gas.
Protect from moisture.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.3; Packaging Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE

storageNote1 : 2° to 8°C