Please login to your online account to display your discounted pricing

4-(Methylthio)phenylboronic acid, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-(Methylsulfanyl)phenylboronic acid
CAS 98546-51-1
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H9BO2S
Formula Weight 168.02
MDL Number MFCD00093410
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC344680010

  SDS
fisher scientific acros
34468-0010
Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 168.02
Physical Form Crystalline Powder
Quantity 1g
Color Off-white
CAS Min % 96.0
CAS Max % 100.0
MDL Number MFCD00093410
Chemical Name or Material 4-(Methylsulfanyl)phenylboronic acid
CAS 98546-51-1
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Packaging Glass Bottle
Molecular Formula C7H9BO2S
Assay Percent Range 97%
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.