Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Methylthio)phenyl isocyanate, 97%

Chemical Identifiers

Chemical Name or Material 4-(Methylthio)phenyl isocyanate
CAS 1632-84-4
Molecular Formula C8H7NOS
Formula Weight 165.22
MDL Number MFCD00013878
UN Number UN2206
Beilstein 1936612
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1226303

 
alfa aesar™
L1226303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1226306

 
alfa aesar™
L1226306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 165.22
CAS 1632-84-4
Assay 97%
Molecular Formula C8H7NOS
MDL Number MFCD00013878
Beilstein 1936612
Quantity 1g
Health Hazard 2 GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
Chemical Name or Material 4-(Methylthio)phenyl isocyanate
UN Number UN2206
Health Hazard 1 Danger
Boiling Point 104° to 105°C (0.5mmHg)
Flash Point 104°C (219°F)
Odor Pungent
Health Hazard 3 GHS P Statement
P260-P280h-P305+P351+P338-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Recommended Storage 2° to 8°C
Safety and Handling

Danger

GHS P Statement
P260-P280h-P305+P351+P338-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C