Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Methylthio)phenyl isocyanate, 97%


Quantity 5g

Manufacturer: alfa aesar™  L1226306

 View more versions of this product

Catalog No. AAL1226306

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 165.22
MDL Number MFCD00013878
CAS 1632-84-4
Molecular Formula C8H7NOS
Chemical Name or Material 4-(Methylthio)phenyl isocyanate
Health Hazard 1 Danger
UN Number UN2206
Health Hazard 3 GHS P Statement
P260-P280h-P305+P351+P338-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Boiling Point 104° to 105°C (0.5mmHg)
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
Beilstein 1936612
Health Hazard 2 GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flash Point 104°C (219°F)
Quantity 5g
Odor Pungent
Recommended Storage 2° to 8°C
Assay 97%

Specifications

Chemical Name or Material 4-(Methylthio)phenyl isocyanate
CAS 1632-84-4
Molecular Formula C8H7NOS
Formula Weight 165.22
MDL Number MFCD00013878
UN Number UN2206
Beilstein 1936612
Accessories

Danger

GHS P Statement
P260-P280h-P305+P351+P338-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C

SDS & Safety Documents