Learn More
4-(Methylthio)phenol, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H2622518
Description
4-(Methylthio)phenol is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-(Methylthio)phenol is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Soluble in water 9.59 g/ l.
Notes
Store in a cool, dry place in tightly closed container. Store away from oxidizing agents.
Specifications
4-(Methylthio)phenol | |
85°C to 87°C | |
Stench | |
MFCD00002351 | |
4-methylthio phenol, 4-methylmercapto phenol, phenol, 4-methylthio, p-methylthio phenol, 4-hydroxythioanisole, 4-methylmercaptophenol, p-hydroxythioanisole, 4-methylsulfanyl phenol, phenol, p-methylthio, p-hydroxyphenyl methyl sulfide | |
QASBCTGZKABPKX-UHFFFAOYSA-N | |
4-methylsulfanylphenol | |
14086 | |
140.21 |
1073-72-9 | |
153°C to 156°C (20 mmHg) | |
C7H8OS | |
50 g | |
Soluble in water 9.59g/ l. | |
CSC1=CC=C(O)C=C1 | |
140.20 | |
CHEBI:38862 | |
98% |
Chemical Identifiers
1073-72-9 | |
140.20 | |
QASBCTGZKABPKX-UHFFFAOYSA-N | |
14086 | |
4-methylsulfanylphenol |
C7H8OS | |
MFCD00002351 | |
4-methylthio phenol, 4-methylmercapto phenol, phenol, 4-methylthio, p-methylthio phenol, 4-hydroxythioanisole, 4-methylmercaptophenol, p-hydroxythioanisole, 4-methylsulfanyl phenol, phenol, p-methylthio, p-hydroxyphenyl methyl sulfide | |
CHEBI:38862 | |
CSC1=CC=C(O)C=C1 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P271-P280-P302+P352-P304+P340-P312-P363-P501c
H312+H332
EINECSNumber : 214-031-4
RTECSNumber : SM1578500
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only