Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Methylthio)benzyl chloride, 99%

Chemical Identifiers

Chemical Name or Material 4-(Methylthio)benzyl chloride
CAS 874-87-3
Molecular Formula C8H9ClS
Formula Weight 172.68
MDL Number MFCD00270118
UN Number UN3265
Synonym 4-(Chloromethyl)thioanisole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1987703

  SDS
alfa aesar™
L1987703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1987706

  SDS
alfa aesar™
L1987706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 172.68
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C8H9ClS
Recommended Storage 2° to 8°C
MDL Number MFCD00270118
Synonym 4-(Chloromethyl)thioanisole
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Chemical Name or Material 4-(Methylthio)benzyl chloride
CAS 874-87-3
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Boiling Point 132°C (22mmHg)
Health Hazard 1 Danger
Quantity 1g
UN Number UN3265
Flash Point >110°C (230°F)
Assay 99%

Applications
4-(Methylthio)benzyl chloride is used as pharmaceutical intermediate.

Solubility
It hydrolyzes in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents and bases.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

2° to 8°C