Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Methylthio)benzonitrile, 98+%

Chemical Identifiers

Chemical Name or Material 4-(Methylthio)benzonitrile
CAS 21382-98-9
Molecular Formula C8H7NS
Formula Weight 149.22
MDL Number MFCD00010381
UN Number UN3439
Beilstein 2079565
Synonym 4-Cyanothioanisole; 4-(Methylmercapto)benzonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0897906

  SDS
alfa aesar™
L0897906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0897914

  SDS
alfa aesar™
L0897914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0897922

  SDS
alfa aesar™
L0897922
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 149.22
MDL Number MFCD00010381
Chemical Name or Material 4-(Methylthio)benzonitrile
Synonym 4-Cyanothioanisole; 4-(Methylmercapto)benzonitrile
Health Hazard 2 GHS H Statement
H311-H302-H332
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Molecular Formula C8H7NS
Health Hazard 3 GHS P Statement
P280h-P309-P310
Wear protective gloves/clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Beilstein 2079565
Health Hazard 1 Danger
CAS 21382-98-9
Assay ≥98%
Boiling Point 272°C
UN Number UN3439
Quantity 5g
Melting Point 59° to 64°C
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

Applications
4-(Methylthio)benzonitrile was used in the preparation of 4-cyanobenzenethiol.

Solubility
Insoluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Safety and Handling

Danger

GHS P Statement
P280h-P309-P310
Wear protective gloves/clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H311-H302-H332
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.