missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Methylthio)benzoic acid, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC163460050

 View more versions of this product

Catalog No. AC163460050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-(Methylthio)benzoic acid
Glass Bottle
192°C to 196°C
13205-48-6
100.0
C8H8O2S
MFCD00002555
4-methylsulfanyl benzoic acid
CSC1=CC=C(C=C1)C(=O)O
168.21
168.22
≥97.5%
Authentic
Yellow to Beige
5g
97.5
98%
CH3SC6H4COOH
10, I, 80
KWHCPERWLHBLOT-UHFFFAOYSA-N
4-methylsulfanylbenzoic acid
83230
Crystalline Powder

Chemical Identifiers

13205-48-6
168.21
KWHCPERWLHBLOT-UHFFFAOYSA-N
83230
CSC1=CC=C(C=C1)C(=O)O
C8H8O2S
MFCD00002555
4-methylsulfanyl benzoic acid
4-methylsulfanylbenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents