Please login to your online account to display your discounted pricing

4-(Methylthio)benzoic acid, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-(Methylthio)benzoic acid
CAS 13205-48-6
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H8O2S
Linear Formula CH3SC6H4COOH
Formula Weight 168.22
MDL Number MFCD00002555
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC163460050

  SDS
thermo fisher scientific
AC163460050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC163460250

  SDS
thermo fisher scientific
AC163460250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 168.22
Physical Form Crystalline Powder
Quantity 5g
Color Light yellow to beige
Melting Point 192° to 196°C
Beilstein 10, I, 80
CAS Max % 100.0
Linear Formula CH3SC6H4COOH
Infrared Spectrum Authentic
Molecular Formula C8H8O2S
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
CAS Min % 97.5
Packaging Glass bottle
Chemical Name or Material 4-(Methylthio)benzoic acid
CAS 13205-48-6
MDL Number MFCD00002555
Assay Percent Range 98%
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.