Learn More
4-(Methylthio)benzoic acid, 97%, Thermo Scientific Chemicals
$66.79 - $848.77
Chemical Identifiers
CAS | 13205-48-6 |
---|---|
Molecular Formula | C8H8O2S |
Molecular Weight (g/mol) | 168.21 |
MDL Number | MFCD00002555 |
InChI Key | KWHCPERWLHBLOT-UHFFFAOYSA-N |
Synonym | 4-methylthio benzoic acid, 4-methylsulfanyl benzoic acid, benzoic acid, 4-methylthio, p-methylthio benzoic acid, p-methiobenzoic acid, benzoic acid, p-methylthio, 4-methylthiobenzoicacid, pubchem10665, acmc-1bqaa |
PubChem CID | 83230 |
IUPAC Name | 4-methylsulfanylbenzoic acid |
SMILES | CSC1=CC=C(C=C1)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2570603
|
Thermo Scientific Chemicals
B2570603 |
1 g |
Each for $66.79
|
|
|||||
AAB2570606
|
Thermo Scientific Chemicals
B2570606 |
5 g |
Each for $209.60
|
|
|||||
AAB2570614
|
Thermo Scientific Chemicals
B2570614 |
25 g |
Each for $848.77
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13205-48-6 | |
168.21 | |
KWHCPERWLHBLOT-UHFFFAOYSA-N | |
83230 | |
CSC1=CC=C(C=C1)C(O)=O |
C8H8O2S | |
MFCD00002555 | |
4-methylthio benzoic acid, 4-methylsulfanyl benzoic acid, benzoic acid, 4-methylthio, p-methylthio benzoic acid, p-methiobenzoic acid, benzoic acid, p-methylthio, 4-methylthiobenzoicacid, pubchem10665, acmc-1bqaa | |
4-methylsulfanylbenzoic acid |
Specifications
13205-48-6 | |
C8H8O2S | |
1 g | |
4-methylthio benzoic acid, 4-methylsulfanyl benzoic acid, benzoic acid, 4-methylthio, p-methylthio benzoic acid, p-methiobenzoic acid, benzoic acid, p-methylthio, 4-methylthiobenzoicacid, pubchem10665, acmc-1bqaa | |
CSC1=CC=C(C=C1)C(O)=O | |
168.21 | |
168.21 | |
4-(Methylthio)benzoic acid |
193°C to 196°C | |
MFCD00002555 | |
2082809 | |
KWHCPERWLHBLOT-UHFFFAOYSA-N | |
4-methylsulfanylbenzoic acid | |
83230 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335-H500
EINECSNumber : 236-171-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only