missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Methylthio)benzaldehyde 97.0+%, TCI America™

Manufacturer:  TCI America M0739250G

 View more versions of this product

Catalog No. M0739250G



Specifications

Specifications

4-(Methylthio)benzaldehyde
Yellow
3446-89-7
MFCD00006948
QRVYABWJVXXOTN-UHFFFAOYSA-N
4-methylsulfanylbenzaldehyde
76985
Clear Liquid at 20°C
153°C
250g
C8H8OS
4-aldehyde thioanisole
CSC1=CC=C(C=C1)C=O
152.211
152.21
≥97.0% (GC)

Chemical Identifiers

3446-89-7
152.211
QRVYABWJVXXOTN-UHFFFAOYSA-N
76985
CSC1=CC=C(C=C1)C=O
C8H8OS
MFCD00006948
4-aldehyde thioanisole
4-methylsulfanylbenzaldehyde
Safety and Handling

Safety and Handling

TSCA : Yes

Documents