missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Methylthio)benzaldehyde, 97%, ACROS Organics™

Manufacturer:  Acros Organics 207150500

 View more versions of this product

Catalog No. AC207150500



Specifications

Specifications

4-(Methylthio)benzaldehyde
>110°C
Glass bottle
1.14
Yellow
50mL
3446-89-7
100.0
C8H8OS
MFCD00006948
4-methylthio benzaldehyde, benzaldehyde, 4-methylthio, 4-methylsulfanyl benzaldehyde, 4-methylmercaptobenzaldehyde, p-methylthio benzaldehyde, p-methylthiobenzaldehyde, 4-methylthio benzaldehyde, unii-1tvq11biy8, 4-aldehyde thioanisole, p-methylmercaptobenzaldehyde
CSC1=CC=C(C=C1)C=O
152.211
152.21
97%
1.1400g/mL
Authentic
1.6452
86.0°C to 90.0°C (1.0mmHg)
6.0°C
97%
96.0
97%
CH3SC6H4CHO
08, I, 533
QRVYABWJVXXOTN-UHFFFAOYSA-N
4-methylsulfanylbenzaldehyde
76985
Liquid

Chemical Identifiers

3446-89-7
152.211
QRVYABWJVXXOTN-UHFFFAOYSA-N
76985
CSC1=CC=C(C=C1)C=O
C8H8OS
MFCD00006948
4-methylthio benzaldehyde, benzaldehyde, 4-methylthio, 4-methylsulfanyl benzaldehyde, 4-methylmercaptobenzaldehyde, p-methylthio benzaldehyde, p-methylthiobenzaldehyde, 4-methylthio benzaldehyde, unii-1tvq11biy8, 4-aldehyde thioanisole, p-methylmercaptobenzaldehyde
4-methylsulfanylbenzaldehyde
Safety and Handling

Safety and Handling

STENCH

EINECSNumber : 222-365-7

TSCA : TSCA

SDS Documents