missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Methylthio)aniline, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 309830050

 View more versions of this product

Catalog No. AC309830050


 

Specifications Description & Specifications

Specifications

4-(Methylthio)aniline
104-96-1
100.0
Authentic
1.6380 to 1.6400 (20°C, 589nm)
Product darkens during storage
98%
CH3SC6H4NH
MFCD00007889
4-Aminothioanisol
98%
97.5
1.119g/mL
Glass bottle
Liquid
≥97.5%
C7H9NS
139.23
Clear orange to brown-red
5g

Chemical Identifiers

104-96-1
100.0
C7H9NS
139.23
4-Aminothioanisol
≥97.5%
97.5
98%
CH3SC6H4NH
MFCD00007889
4-(Methylthio)aniline, 98%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 203-256-3

RTECSNumber : BY6302000

TSCA : TSCA