missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Methylthio)-6-oxo-2-phenyl-1,6-dihydropyrimidine-5-carbonitrile, ≥97%, Maybridge

Chemical Identifiers

CAS 15908-64-2
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97+%
Molecular Formula C12H9N3OS
Formula Weight 243.29
MDL Number MFCD00067844
Chemical Name or Material 4-(Methylthio)-6-oxo-2-phenyl-1, 6-dihydropyrimidine-5-carbonitrile
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
BTB08867DA SDS Fisher Scientific Company
BTB08867DA
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

15908-64-2
100.0
C12H9N3OS
MFCD00067844
97%
96.5
97+%
243.29
4-(Methylthio)-6-oxo-2-phenyl-1, 6-dihydropyrimidine-5-carbonitrile

Specifications

243.29
100.0
C12H9N3OS
96.5
4-(Methylthio)-6-oxo-2-phenyl-1, 6-dihydropyrimidine-5-carbonitrile
1g
97+%
MFCD00067844
15908-64-2
97%

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning