missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methylthiazole 98.0+%, TCI America™

$28.80 - $250.13

Chemical Identifiers

CAS 693-95-8
Molecular Formula C4H5NS
Molecular Weight (g/mol) 99.151
MDL Number MFCD00005340
InChI Key QMHIMXFNBOYPND-UHFFFAOYSA-N
Synonym 4-methyithiazole
PubChem CID 12748
ChEBI CHEBI:35626
IUPAC Name 4-methyl-1,3-thiazole
SMILES CC1=CSC=N1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M08945G View Documents Tci America
M08945G
5g Each for $28.80
 
M089425G View Documents Tci America
M089425G
25g Each for $83.38
 
M0894100G View Documents Tci America
M0894100G
100g Each for $250.13
 
Specifications

Chemical Identifiers

693-95-8
99.151
QMHIMXFNBOYPND-UHFFFAOYSA-N
12748
4-methyl-1,3-thiazole
C4H5NS
MFCD00005340
4-methyithiazole
CHEBI:35626
CC1=CSC=N1

Specifications

134°C
5g
C4H5NS
1993
QMHIMXFNBOYPND-UHFFFAOYSA-N
4-methyl-1,3-thiazole
12748
99.15
≥98.0% (GC)
Yellow
693-95-8
MFCD00005340
4-methyithiazole
CC1=CSC=N1
99.151
CHEBI:35626
Clear Liquid at 20°C
4-Methylthiazole
Safety and Handling

Safety and Handling

RTECSNumber : XJ5096000

TSCA : Yes