Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methylthiazole, 99%

$26.49 - $26.49

Chemical Identifiers

Chemical Name or Material 4-Methylthiazole
CAS 693-95-8
Molecular Formula C4H5NS
Formula Weight 99.16
MDL Number MFCD00005340
UN Number UN1993
Beilstein 105228
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1542806

  SDS
alfa aesar™
A1542806
5g Each for $26.49

AAA1542814

  SDS
alfa aesar™
A1542814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 99.16
Chemical Name or Material 4-Methylthiazole
TSCA TSCA
Assay 99%
Molecular Formula C4H5NS
MDL Number MFCD00005340
CAS 693-95-8
UN Number UN1993
Beilstein 105228
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H226-H315-H319-H335
Toxic if swallowed.
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
RTECS Number XJ5096000
Flash Point 32°C (89°F)
Odor Characteristic
Boiling Point 133° to 134°C
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H226-H315-H319-H335
Toxic if swallowed.
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.