Please login to your online account to display your discounted pricing

4-Methylthiazole-5-carboxylic acid, 97%, Acros Organics


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC396581000

 View more versions of this product

Catalog No. AC396581000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material 4-Methylthiazole-5-carboxylic acid
Quantity 100g
CAS 20485-41-0
Packaging Glass bottle
Assay Percent Range 97%
Molecular Formula C5H5NO2S
Formula Weight 143.17

Specifications

Chemical Name or Material 4-Methylthiazole-5-carboxylic acid
Assay Percent Range 97%
Molecular Formula C5H5NO2S
Formula Weight 143.17