missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Methylthiazole-5-carboxaldehyde, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 82294-70-0 |
---|---|
Molecular Formula | C5H5NOS |
Molecular Weight (g/mol) | 127.17 |
MDL Number | MFCD07368277 |
InChI Key | JJVIEMFQPALZOZ-UHFFFAOYSA-N |
Synonym | 4-methyl-5-formylthiazole, 4-methyl-5-thiazole carboxaldehyde, 4-methyl-5-thiazolecarbaldehyde, 4-methyl-5-thiazolecarboxaldehyde, 4-methyl-5-thiazolyl aldehyde, 4-methylthiazole-5-aldehyde, 4-methylthiazole-5-carbaldehyde, 4-methylthiazole-5-carboxaldehyde, 5-formyl-4-methylthiazole, 5-thiazolecarboxaldehyde, 4-methyl |
PubChem CID | 581339 |
IUPAC Name | 4-methyl-1,3-thiazole-5-carbaldehyde |
SMILES | CC1=C(SC=N1)C=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC429970010
|
Thermo Fisher Scientific
AC429970010 |
1g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
82294-70-0 | |
127.17 | |
JJVIEMFQPALZOZ-UHFFFAOYSA-N | |
581339 | |
CC1=C(SC=N1)C=O |
C5H5NOS | |
MFCD07368277 | |
4-methyl-5-formylthiazole, 4-methyl-5-thiazole carboxaldehyde, 4-methyl-5-thiazolecarbaldehyde, 4-methyl-5-thiazolecarboxaldehyde, 4-methyl-5-thiazolyl aldehyde, 4-methylthiazole-5-aldehyde, 4-methylthiazole-5-carbaldehyde, 4-methylthiazole-5-carboxaldehyde, 5-formyl-4-methylthiazole, 5-thiazolecarboxaldehyde, 4-methyl | |
4-methyl-1,3-thiazole-5-carbaldehyde |
Specifications
82294-70-0 | |
100.0 | |
96% min. (GC) | |
MFCD07368277 | |
4-methyl-5-formylthiazole, 4-methyl-5-thiazole carboxaldehyde, 4-methyl-5-thiazolecarbaldehyde, 4-methyl-5-thiazolecarboxaldehyde, 4-methyl-5-thiazolyl aldehyde, 4-methylthiazole-5-aldehyde, 4-methylthiazole-5-carbaldehyde, 4-methylthiazole-5-carboxaldehyde, 5-formyl-4-methylthiazole, 5-thiazolecarboxaldehyde, 4-methyl | |
JJVIEMFQPALZOZ-UHFFFAOYSA-N | |
4-methyl-1,3-thiazole-5-carbaldehyde | |
581339 | |
97% |
96.0 | |
74.0°C to 76.0°C | |
C5H5NOS | |
Glass bottle | |
1g | |
CC1=C(SC=N1)C=O | |
127.17 | |
127.17 | |
4-Methylthiazole-5-carboxaldehyde |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
GHS Signal Word: Danger
RUO – Research Use Only