Please login to your online account to display your discounted pricing

4-Methylsulphonylphenol, 95%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC424880100

 View more versions of this product

Catalog No. AC424880100

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥94%
Formula Weight 172.2
Physical Form Powder
Quantity 10g
Color Tan to pink-brown
Melting Point 90° to 95°C
Water 5% Max.
CAS Min % 94.0
CAS Max % 100.0
Chemical Name or Material 4-Methylsulphonylphenol
Assay Percent Range 95%
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula HOC6H4SO2CH3
Molecular Formula C7H8O3S
CAS 14763-60-1
MDL Number MFCD00047806
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Specifications

Chemical Name or Material 4-Methylsulphonylphenol
CAS 14763-60-1
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C7H8O3S
Linear Formula HOC6H4SO2CH3
Formula Weight 172.2
MDL Number MFCD00047806
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents