Please login to your online account to display your discounted pricing

4-Methylsulphonylphenol, 95%, ACROS Organics™

$306.81 - $306.81

Chemical Identifiers

Chemical Name or Material 4-Methylsulphonylphenol
CAS 14763-60-1
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C7H8O3S
Linear Formula HOC6H4SO2CH3
Formula Weight 172.2
MDL Number MFCD00047806
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC424880100

  SDS
thermo fisher scientific
AC424880100
Glass bottle 10g Each for $306.81

AC424880025

  SDS
fisher scientific acros
424880025
Glass bottle 2.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Physical Form Powder
Formula Weight 172.2
Quantity 10g
Color Tan to pink-brown
Water 5% Max.
Melting Point 90° to 95°C
CAS Min % 94.0
CAS Max % 100.0
Chemical Name or Material 4-Methylsulphonylphenol
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 95%
Molecular Formula C7H8O3S
Linear Formula HOC6H4SO2CH3
MDL Number MFCD00047806
CAS 14763-60-1
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.