Please login to your online account to display your discounted pricing

4-Methylsulphonylbenzyl bromide, Tech., ACROS Organics™


Quantity 2.5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  357580025

 View more versions of this product

Catalog No. AC357580025

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥60%
Formula Weight 249.13
Purity Grade Technical Grade
Physical Form Crystalline Powder
Quantity 2.5g
Color Beige to brown
Beilstein 06, IV, 2210
Packaging Glass bottle
Molecular Formula C8H9BrO2S
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Infrared Spectrum Authentic
Chemical Name or Material 4-Methylsulphonylbenzyl bromide
CAS 53606-06-7
MDL Number MFCD00185849
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4-Methylsulphonylbenzyl bromide
CAS 53606-06-7
Purity Grade Technical Grade
Assay ≥60%
Molecular Formula C8H9BrO2S
Formula Weight 249.13
MDL Number MFCD00185849
Accessories

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.

SDS & Safety Documents