missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methylsulphonylbenzoic acid, 95%, ACROS Organics™

Chemical Identifiers

CAS 4052-30-6
Molecular Formula C8H8O4S
Molecular Weight (g/mol) 200.22
MDL Number MFCD00007564
InChI Key AJBWNNKDUMXZLM-UHFFFAOYSA-N
Synonym 4-methanesulfonyl benzoic acid, 4-methanesulfonyl-benzoic acid, 4-methanesulfonylbenzoic acid, 4-methylsulfonyl benzoic acid, 4-methylsulphonyl benzoic acid, 4-methylsulphonylbenzoic acid, benzoic acid, 4-methylsulfonyl, p-methylsulfonylbenzoic acid, p-methylsulphonyl benzoic acid
PubChem CID 77670
IUPAC Name 4-methylsulfonylbenzoic acid
SMILES CS(=O)(=O)C1=CC=C(C=C1)C(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC424920025
SDS View Documents
Thermo Fisher Scientific
424920025
2.5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

4052-30-6
200.22
AJBWNNKDUMXZLM-UHFFFAOYSA-N
77670
CS(=O)(=O)C1=CC=C(C=C1)C(=O)O
C8H8O4S
MFCD00007564
4-methanesulfonyl benzoic acid, 4-methanesulfonyl-benzoic acid, 4-methanesulfonylbenzoic acid, 4-methylsulfonyl benzoic acid, 4-methylsulphonyl benzoic acid, 4-methylsulphonylbenzoic acid, benzoic acid, 4-methylsulfonyl, p-methylsulfonylbenzoic acid, p-methylsulphonyl benzoic acid
4-methylsulfonylbenzoic acid

Specifications

Authentic
Glass bottle
270°C to 275°C
4052-30-6
100.0
C8H8O4S
MFCD00007564
Solubility in water: slight
CS(=O)(=O)C1=CC=C(C=C1)C(=O)O
200.22
200.22
95%
5% max.
White
2.5g
94.0
94% min. (Acid)
HOCC(O)C6H4SO2CH3
4-methanesulfonyl benzoic acid, 4-methanesulfonyl-benzoic acid, 4-methanesulfonylbenzoic acid, 4-methylsulfonyl benzoic acid, 4-methylsulphonyl benzoic acid, 4-methylsulphonylbenzoic acid, benzoic acid, 4-methylsulfonyl, p-methylsulfonylbenzoic acid, p-methylsulphonyl benzoic acid
AJBWNNKDUMXZLM-UHFFFAOYSA-N
4-methylsulfonylbenzoic acid
77670
Powder
4-Methylsulphonylbenzoic acid
Safety and Handling

Safety and Handling

EINECSNumber : 223-756-5

SDS