Please login to your online account to display your discounted pricing

4-Methylsulphonylbenzoic acid, 95%, ACROS Organics™


Quantity 2.5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  424920025

 View more versions of this product

Catalog No. AC424920025

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Powder
Formula Weight 200.22
Quantity 2.5g
Color White
Melting Point 270° to 275°C
CAS Min % 94.0
CAS Max % 100.0
Chemical Name or Material 4-Methylsulphonylbenzoic acid
Loss on Drying 5% max.
Packaging Glass bottle
Assay Percent Range 95%
Molecular Formula C8 H8 O4 S
CAS 4052-30-6
MDL Number MFCD00007564
Infrared Spectrum Authentic
Linear Formula HOCC(O)C6H4SO2CH3

Specifications

Chemical Name or Material 4-Methylsulphonylbenzoic acid
CAS 4052-30-6
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C8 H8 O4 S
Linear Formula HOCC(O)C6H4SO2CH3
Formula Weight 200.22
MDL Number MFCD00007564