Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Methylsulfonylamino)benzonitrile, 97%

Chemical Identifiers

Chemical Name or Material 4-(Methylsulfonylamino)benzonitrile
CAS 36268-67-4
Molecular Formula C8H8N2O2S
Formula Weight 196.23
MDL Number MFCD00204724
UN Number UN3439
Synonym 4-(Methanesulfonylamino)benzonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2043903

  SDS
alfa aesar™
B2043903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2043906

  SDS
alfa aesar™
B2043906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 196.23
Melting Point 192° to 196°C
Molecular Formula C8H8N2O2S
Chemical Name or Material 4-(Methylsulfonylamino)benzonitrile
UN Number UN3439
CAS 36268-67-4
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Quantity 1g
Synonym 4-(Methanesulfonylamino)benzonitrile
Assay 97%
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
MDL Number MFCD00204724
Safety and Handling

Danger

GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.