Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Methylsulfonyl)phenol, 97%

$50.34 - $315.55

Chemical Identifiers

Chemical Name or Material 4-(Methylsulfonyl)phenol
CAS 14763-60-1
Molecular Formula C7H8O3S
Formula Weight 172.2
MDL Number MFCD00047806
Synonym 4-Hydroxyphenyl methyl sulfone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3293503

  SDS
alfa aesar™
H3293503
1g Each for $50.34

AAH3293509

  SDS
alfa aesar™
H3293509
10g Each for $315.55
Description & Specifications

Specifications

Formula Weight 172.2
Chemical Name or Material 4-(Methylsulfonyl)phenol
CAS 14763-60-1
Health Hazard 1 Warning
Melting Point 90° to 95°C
Quantity 1g
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RTECS Number SM1530000
Assay 97%
Molecular Formula C7H8O3S
MDL Number MFCD00047806
Synonym 4-Hydroxyphenyl methyl sulfone
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.