Please login to your online account to display your discounted pricing

4-(Methylsulfonyl)benzenesulfonyl chloride, 95%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC424970100

 View more versions of this product

Catalog No. AC424970100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94% (ex Cl)
Physical Form Crystalline Powder
Formula Weight 254.71
Quantity 10g
Color Yellow to salmon-pink
Melting Point 158° to 163°C
CAS Min % 94.0
CAS Max % 100.0
Chemical Name or Material 4-(Methylsulfonyl)benzenesulfonyl chloride
Infrared Spectrum Authentic
Assay Percent Range 95%
Molecular Formula C7H7ClO4S2
CAS 82964-91-8
Loss on Drying 5% max.
Packaging Glass bottle
Linear Formula ClO2SC6H4SO2CH3
MDL Number MFCD00216486

Specifications

Chemical Name or Material 4-(Methylsulfonyl)benzenesulfonyl chloride
CAS 82964-91-8
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94% (ex Cl)
Assay Percent Range 95%
Molecular Formula C7H7ClO4S2
Linear Formula ClO2SC6H4SO2CH3
Formula Weight 254.71
MDL Number MFCD00216486