Please login to your online account to display your discounted pricing

4-(Methylsulfonyl)benzenesulfonyl chloride, 95%, ACROS Organics™

$113.49 - $113.49

Chemical Identifiers

Chemical Name or Material 4-(Methylsulfonyl)benzenesulfonyl chloride
CAS 82964-91-8
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94% (ex Cl)
Assay Percent Range 95%
Molecular Formula C7H7ClO4S2
Linear Formula ClO2SC6H4SO2CH3
Formula Weight 254.71
MDL Number MFCD00216486
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC424970025

  SDS
thermo fisher scientific
424970025
Glass bottle 2.5g Each for $113.49
Description & Specifications

Specifications

Assay ≥94% (ex Cl)
Physical Form Crystalline Powder
Formula Weight 254.71
Quantity 2.5g
Color Yellow to salmon-pink
Melting Point 158° to 163°C
CAS Min % 94.0
CAS Max % 100.0
Linear Formula ClO2SC6H4SO2CH3
Infrared Spectrum Authentic
Loss on Drying 5% max.
CAS 82964-91-8
Molecular Formula C7H7ClO4S2
Chemical Name or Material 4-(Methylsulfonyl)benzenesulfonyl chloride
Assay Percent Range 95%
MDL Number MFCD00216486
Packaging Glass bottle