Please login to your online account to display your discounted pricing

4-(Methylsulfonyl)benzenesulfonyl chloride, 95%, ACROS Organics™


Quantity 2.5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  424970025

 View more versions of this product

Catalog No. AC424970025

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥94% (ex Cl)
Physical Form Crystalline Powder
Formula Weight 254.71
Quantity 2.5g
Color Yellow to salmon-pink
Melting Point 158° to 163°C
CAS Min % 94.0
CAS Max % 100.0
Linear Formula ClO2SC6H4SO2CH3
Infrared Spectrum Authentic
Loss on Drying 5% max.
CAS 82964-91-8
Molecular Formula C7H7ClO4S2
Chemical Name or Material 4-(Methylsulfonyl)benzenesulfonyl chloride
Assay Percent Range 95%
MDL Number MFCD00216486
Packaging Glass bottle

Specifications

Chemical Name or Material 4-(Methylsulfonyl)benzenesulfonyl chloride
CAS 82964-91-8
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94% (ex Cl)
Assay Percent Range 95%
Molecular Formula C7H7ClO4S2
Linear Formula ClO2SC6H4SO2CH3
Formula Weight 254.71
MDL Number MFCD00216486