Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Methylsulfonyl)benzeneboronic acid, 98%

$46.72 - $500.24

Chemical Identifiers

Chemical Name or Material 4-(Methylsulfonyl)benzeneboronic acid
CAS 149104-88-1
Molecular Formula C7H9BO4S
Formula Weight 200.02
MDL Number MFCD01630820
Synonym 4-(Methanesulfonyl)benzeneboronic acid; 4-(Methylsulfonyl)phenylboronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1772003

  SDS
alfa aesar™
L1772003
1g Each for $133.04

AAL1772006

  SDS
alfa aesar™
L1772006
5g Each for $500.24

AAL17720MD

  SDS
alfa aesar™
L17720MD
250mg Each for $46.72
Description & Specifications

Specifications

Formula Weight 200.02
Chemical Name or Material 4-(Methylsulfonyl)benzeneboronic acid
Assay 98%
MDL Number MFCD01630820
Synonym 4-(Methanesulfonyl)benzeneboronic acid; 4-(Methylsulfonyl)phenylboronic acid
Health Hazard 1 Warning
Melting Point ≈200°C
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C7H9BO4S
CAS 149104-88-1
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.