missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-(Methylsulfonyl)acetophenone, 97+%

$40.03 - $128.09

Chemical Identifiers

CAS 10297-73-1
Molecular Formula C9H10O3S
Molecular Weight (g/mol) 198.236
MDL Number MFCD00025072
InChI Key KAVZYDHKJNABPC-UHFFFAOYSA-N
Synonym 1-4-methanesulfonyl-phenyl-ethanone
PubChem CID 82529
IUPAC Name 1-(4-methylsulfonylphenyl)ethanone
SMILES CC(=O)C1=CC=C(C=C1)S(=O)(=O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5407203 View Documents Alfa Aesar™
H5407203
1g Each for $40.03
 
AAH5407206 View Documents Alfa Aesar™
H5407206
5g Each for $128.09
 
Specifications

Chemical Identifiers

10297-73-1
198.236
KAVZYDHKJNABPC-UHFFFAOYSA-N
82529
CC(=O)C1=CC=C(C=C1)S(=O)(=O)C
C9H10O3S
MFCD00025072
1-4-methanesulfonyl-phenyl-ethanone
1-(4-methylsulfonylphenyl)ethanone

Specifications

126°C to 129°C
10297-73-1
MFCD00025072
KAVZYDHKJNABPC-UHFFFAOYSA-N
1-(4-methylsulfonylphenyl)ethanone
82529
≥97%
1g
C9H10O3S
1-4-methanesulfonyl-phenyl-ethanone
CC(=O)C1=CC=C(C=C1)S(=O)(=O)C
198.236
198.24
4'-(Methylsulfonyl)acetophenone
Safety and Handling

Safety and Handling

EINECSNumber : 233-672-0

TSCA : No

Recommended Storage : Ambient temperatures