missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-(Methylsulfonyl)acetanilide, 96%

$73.96 - $209.80

Chemical Identifiers

CAS 22821-80-3
Molecular Formula C9H11NO3S
Molecular Weight (g/mol) 213.251
MDL Number MFCD00025068
InChI Key SJYUABJSWXGSAO-UHFFFAOYSA-N
Synonym 4'-methylsulfonyl acetanilide, 4'-methylsulfonylacetanilide, 4-methylsulphonyl-acetanilide, acmc-1cqab, n-4-methanesulfonyl-phenyl-acetamide, n-4-methanesulfonylphenyl acetamide, n-4-methylsulfonyl phenyl acetamide, n-4-methylsulfonylphenyl acetamide, p-methylsulfonyl acetanilide
PubChem CID 257586
IUPAC Name N-(4-methylsulfonylphenyl)acetamide
SMILES CC(=O)NC1=CC=C(C=C1)S(=O)(=O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2692206
View Documents
Alfa Aesar™
H2692206
5g
Each for $73.96
 
AAH2692214
View Documents
Alfa Aesar™
H2692214
25g
Each for $209.80
 
Description

Description

Specifications

Chemical Identifiers

22821-80-3
213.251
SJYUABJSWXGSAO-UHFFFAOYSA-N
257586
CC(=O)NC1=CC=C(C=C1)S(=O)(=O)C
C9H11NO3S
MFCD00025068
4'-methylsulfonyl acetanilide, 4'-methylsulfonylacetanilide, 4-methylsulphonyl-acetanilide, acmc-1cqab, n-4-methanesulfonyl-phenyl-acetamide, n-4-methanesulfonylphenyl acetamide, n-4-methylsulfonyl phenyl acetamide, n-4-methylsulfonylphenyl acetamide, p-methylsulfonyl acetanilide
N-(4-methylsulfonylphenyl)acetamide

Specifications

5g
C9H11NO3S
4'-methylsulfonyl acetanilide, 4'-methylsulfonylacetanilide, 4-methylsulphonyl-acetanilide, acmc-1cqab, n-4-methanesulfonyl-phenyl-acetamide, n-4-methanesulfonylphenyl acetamide, n-4-methylsulfonyl phenyl acetamide, n-4-methylsulfonylphenyl acetamide, p-methylsulfonyl acetanilide
CC(=O)NC1=CC=C(C=C1)S(=O)(=O)C
213.251
213.26
4'-(Methylsulfonyl)acetanilide
22821-80-3
MFCD00025068
SJYUABJSWXGSAO-UHFFFAOYSA-N
N-(4-methylsulfonylphenyl)acetamide
257586
96%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Product Certifications