missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-(Methylsulfonyl)acetanilide, 96%

Manufacturer:  Alfa Aesar™ H2692206

 View more versions of this product

Catalog No. AAH2692206



Description

Description

Specifications

Specifications

4'-(Methylsulfonyl)acetanilide
22821-80-3
MFCD00025068
SJYUABJSWXGSAO-UHFFFAOYSA-N
N-(4-methylsulfonylphenyl)acetamide
257586
96%
5g
C9H11NO3S
4'-methylsulfonyl acetanilide, 4'-methylsulfonylacetanilide, 4-methylsulphonyl-acetanilide, acmc-1cqab, n-4-methanesulfonyl-phenyl-acetamide, n-4-methanesulfonylphenyl acetamide, n-4-methylsulfonyl phenyl acetamide, n-4-methylsulfonylphenyl acetamide, p-methylsulfonyl acetanilide
CC(=O)NC1=CC=C(C=C1)S(=O)(=O)C
213.251
213.26

Chemical Identifiers

22821-80-3
213.251
SJYUABJSWXGSAO-UHFFFAOYSA-N
257586
CC(=O)NC1=CC=C(C=C1)S(=O)(=O)C
C9H11NO3S
MFCD00025068
4'-methylsulfonyl acetanilide, 4'-methylsulfonylacetanilide, 4-methylsulphonyl-acetanilide, acmc-1cqab, n-4-methanesulfonyl-phenyl-acetamide, n-4-methanesulfonylphenyl acetamide, n-4-methylsulfonyl phenyl acetamide, n-4-methylsulfonylphenyl acetamide, p-methylsulfonyl acetanilide
N-(4-methylsulfonylphenyl)acetamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents
Product Certifications