Learn More
4-Methylsalicylic acid, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 223540250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4-Methylsalicylic acid | |
97.5 | |
Authentic | |
Glass bottle | |
CH3C6H3(OH)CO2H | |
25g | |
15, 2569 | |
NJESAXZANHETJV-UHFFFAOYSA-N | |
2-hydroxy-4-methylbenzoic acid | |
5788 | |
152.15 |
50-85-1 | |
100.0 | |
98% | |
C8H8O3 | |
MFCD00002454 | |
10, 233 | |
4-methylsalicylic acid, m-cresotic acid, 2,4-cresotic acid, m-cresotinic acid, 2-hydroxy-p-toluic acid, m-homosalicylic acid, 4-methyl-2-hydroxybenzoic acid, gamma-cresotic acid, benzoic acid, 2-hydroxy-4-methyl, 4-methylsalicylate | |
CC1=CC(=C(C=C1)C(=O)O)O | |
152.15 | |
CHEBI:20450 | |
98% |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
EINECSNumber : 200-068-3
RUO â Research Use Only