Learn More
4-Methylpyrimidine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1317014
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Methylpyrimidine | |
1.03 | |
40°C (104°F) | |
1.496 | |
25 g | |
105781 | |
LVILGAOSPDLNRM-UHFFFAOYSA-N | |
4-methylpyrimidine | |
18922 | |
98% |
3438-46-8 | |
141°C to 142°C | |
C5H6N2 | |
MFCD00006115 | |
UN1993 | |
pyrimidine, 4-methyl, 4-methyl-pyrimidine, 4-methyl pyrimidine, pubchem20803, 6-methylpyrimidine, 4-methylpyrimidine, ksc226a9b, tpc-h013, 4-methyl-1,3-diazine, pyrimidine, 4-methyl-6ci,7ci,8ci,9ci | |
CC1=NC=NC=C1 | |
94.117 | |
94.12 |
Chemical Identifiers
3438-46-8 | |
94.117 | |
LVILGAOSPDLNRM-UHFFFAOYSA-N | |
18922 | |
CC1=NC=NC=C1 |
C5H6N2 | |
MFCD00006115 | |
pyrimidine, 4-methyl, 4-methyl-pyrimidine, 4-methyl pyrimidine, pubchem20803, 6-methylpyrimidine, 4-methylpyrimidine, ksc226a9b, tpc-h013, 4-methyl-1,3-diazine, pyrimidine, 4-methyl-6ci,7ci,8ci,9ci | |
4-methylpyrimidine |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapour.
P201-P202-P210-P233-P240-P241-P242-P243-P281-P303+P361+P353-P308+P313-P370+P378q-P501c
H226-H360D
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 222-344-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only