missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methylpyrimidine, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC155770500

 View more versions of this product

Catalog No. AC155770500

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥96.0%
Liquid
Clear colorless to slightly yellow
23, 92
96.0
C5H6N2
4-Methylpyrimidine
MFCD00006115
1.4950 to 1.4970 (20°C, 589nm)
94.12
50g
1.0310g/mL
100.0
Glass bottle
97%
3438-46-8
Authentic

Chemical Identifiers

3438-46-8
100.0
C5H6N2
MFCD00006115
≥96.0%
96.0
97%
94.12
4-Methylpyrimidine

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May damage the unborn child.
Flammable liquid and vapor.

GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger