Please login to your online account to display your discounted pricing

4-Methylpyrimidine, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Methylpyrimidine
CAS 3438-46-8
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C5H6N2
Formula Weight 94.12
MDL Number MFCD00006115
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC155770500

  SDS
thermo fisher scientific
AC155770500
Glass bottle 50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid
Formula Weight 94.12
Quantity 50g
Color Clear colorless to slightly yellow
Density 1.0310g/mL
Beilstein 23, 92
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 4-Methylpyrimidine
Packaging Glass bottle
Assay Percent Range 97%
Molecular Formula C5H6N2
MDL Number MFCD00006115
CAS 3438-46-8
Infrared Spectrum Authentic
Refractive Index 1.4950 to 1.4970 (20°C, 589nm)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
May damage the unborn child.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

Danger

GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May damage the unborn child.
Flammable liquid and vapor.