Please login to your online account to display your discounted pricing

4-Methylpyridine-3-carboxylic acid, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Methylpyridine-3-carboxylic acid
CAS 3222-50-2
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H7NO2
Formula Weight 137.14
MDL Number MFCD00128869
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC429420050

  SDS
thermo fisher scientific
AC429420050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC429420010

  SDS
thermo fisher scientific
AC429420010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥96.0%
Formula Weight 137.14
Quantity 5g
Color White to tan
Melting Point 212° to 220°C
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 4-Methylpyridine-3-carboxylic acid
Assay Percent Range 97%
CAS 3222-50-2
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C7H7NO2
MDL Number MFCD00128869
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.