Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methylpyridazine, 99%

$52.28 - $170.27

Chemical Identifiers

Chemical Name or Material 4-Methylpyridazine
CAS 1120-88-3
Molecular Formula C5H6N2
Formula Weight 94.12
MDL Number MFCD00011594
Beilstein 105689
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL09749MC

  SDS
alfa aesar™
L09749MC
100mg Each for $52.28

AAL09749ME

  SDS
alfa aesar™
L09749ME
500mg Each for $170.27
Description & Specifications

Specifications

Formula Weight 94.12
Chemical Name or Material 4-Methylpyridazine
Molecular Formula C5H6N2
Assay 99%
Boiling Point 98° to 100°C (11mmHg)
Health Hazard 1 Warning
CAS 1120-88-3
Beilstein 105689
MDL Number MFCD00011594
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 105°C (221°F)
Quantity 100mg
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.